Assay Details
| Gene Symbol: eppl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028175.6 | NP_001023346.1 | 3 - 4 | 397 | 67 | |
| NR_131583.1 | 3 - 4 | 390 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eppl-1| Gene Symbol: eppl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028175.6 | NP_001023346.1 | 3 - 4 | 397 | 67 | |
| NR_131583.1 | 3 - 4 | 390 | 67 | |||
Set Membership: |
|