Assay Details
| Gene Symbol: dgk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069075.6 | NP_501476.2 | 11 - 12 | 2180 | 70 | |
| NM_069076.5 | NP_501477.2 | 11 - 12 | 2181 | 70 | ||
More Information
Set Membership: |
|

dgk-4
Genomic Map