Assay Details
| Gene Symbol: dgk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069075.6 | NP_501476.2 | 16 - 17 | 3474 | 88 | |
| NM_069076.5 | NP_501477.2 | 15 - 16 | 3322 | 88 | ||
More Information
Set Membership: |
|

dgk-4
Genomic Map