Assay Details
| Gene Symbol: cdh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307097.4 | NP_001294026.1 | 10 - 11 | 2113 | 112 | |
| NM_001356929.4 | NP_001343640.1 | 12 | 2545 | 112 | ||
| NM_001356930.4 | NP_001343703.1 | 10 | 2338 | 112 | ||
| NM_069086.6 | NP_501487.1 | 10 - 11 | 2034 | 112 | ||
More Information
Set Membership: |
|

cdh-5
Genomic Map