Assay Details
Gene Symbol: cdh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307097.4 | NP_001294026.1 | 2 - 3 | 432 | 71 | |
NM_001356929.4 | NP_001343640.1 | 3 | 435 | 71 | ||
NM_001356930.4 | NP_001343703.1 | 1 | 228 | 71 | ||
NM_069086.6 | NP_501487.1 | 2 - 3 | 353 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
