Assay Details
Gene Symbol: cdh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307097.4 | NP_001294026.1 | 7 - 8 | 1554 | 153 | |
NM_001356929.4 | NP_001343640.1 | 7 - 8 | 1557 | 153 | ||
NM_001356930.4 | NP_001343703.1 | 6 | 1350 | 153 | ||
NM_069086.6 | NP_501487.1 | 7 - 8 | 1475 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
