Assay Details
| Gene Symbol: cdh-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356929.4 | NP_001343640.1 | 22 | 4482 | 83 | |
| NM_069086.6 | NP_501487.1 | 20 - 21 | 3971 | 83 | ||
More Information
Set Membership: |
|

cdh-5
Genomic Map