Assay Details
Gene Symbol: ints-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307283.3 | NP_001294212.1 | 1 - 2 | 495 | 90 | |
NM_001307284.2 | NP_001294213.1 | 1 - 2 | 158 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
