Assay Details
| Gene Symbol: F21D5.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069099.7 | NP_501500.1 | 4 - 5 | 992 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F21D5.1| Gene Symbol: F21D5.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069099.7 | NP_501500.1 | 4 - 5 | 992 | 69 | |
Set Membership: |
|