Assay Details
Gene Symbol: rap-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069148.5 | NP_501549.1 | 3 - 4 | 297 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
