Assay Details
| Gene Symbol: mrpl-42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069179.9 | NP_501580.3 | 3 - 4 | 474 | 99 | |
| Gene Symbol: icln-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383158.1 | 7 - 8 | 1348 | 99 | ||
| Gene Symbol: lpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348646.4 | NP_001335548.1 | 3 | 497 | 99 | |
More Information
Set Membership: |
|

mrpl-42
Genomic Map