Assay Details
| Gene Symbol: nmy-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069219.6 | NP_501620.2 | 9 - 10 | 1443 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nmy-3| Gene Symbol: nmy-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069219.6 | NP_501620.2 | 9 - 10 | 1443 | 112 | |
Set Membership: |
|