Assay Details
| Gene Symbol: aat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069306.7 | NP_501707.1 | 5 - 6 | 1159 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aat-1| Gene Symbol: aat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069306.7 | NP_501707.1 | 5 - 6 | 1159 | 104 | |
Set Membership: |
|