Assay Details
| Gene Symbol: mca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028255.4 | NP_001023426.1 | 6 - 7 | 2028 | 70 | |
| NM_001028256.7 | NP_001023427.1 | 6 - 7 | 2117 | 70 | ||
| NM_001268480.3 | NP_001255409.1 | 3 - 4 | 1538 | 70 | ||
| NM_069308.6 | NP_501709.2 | 6 - 7 | 2118 | 70 | ||
| GenBank mRNA | AJ223616.1 | - | 6 - 7 | 2050 | 70 | |
| AY438644.1 | - | 6 - 7 | 2118 | 70 | ||
| AY438645.1 | - | 6 - 7 | 2090 | 70 | ||
More Information
Set Membership: |
|

mca-1
Genomic Map