Assay Details
| Gene Symbol: cav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268492.3 | NP_001255421.1 | 1 - 2 | 293 | 92 | |
| GenBank mRNA | U66405.1 | - | 1 - 2 | 271 | 92 | |
More Information
Set Membership: |
|

cav-1
Genomic Map