Assay Details
| Gene Symbol: sna-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069343.6 | NP_501744.1 | 5 - 6 | 1126 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sna-2| Gene Symbol: sna-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069343.6 | NP_501744.1 | 5 - 6 | 1126 | 72 | |
Set Membership: |
|