Assay Details
| Gene Symbol: mtr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069356.6 | NP_501757.1 | 12 - 13 | 2036 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtr-4| Gene Symbol: mtr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069356.6 | NP_501757.1 | 12 - 13 | 2036 | 79 | |
Set Membership: |
|