Assay Details
Gene Symbol: gon-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268519.3 | NP_001255448.1 | 4 - 5 | 576 | 69 | |
NM_001372913.1 | NP_001359558.1 | 7 - 8 | 1080 | 69 | ||
NR_056474.1 | 7 - 8 | 1076 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
