Assay Details
| Gene Symbol: uba-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038315.8 | NP_001033404.1 | 2 - 3 | 357 | 75 | |
| NM_001380424.1 | NP_001367213.1 | 2 - 3 | 332 | 75 | ||
| NM_001383176.1 | 2 - 3 | 270 | 75 | |||
More Information
Set Membership: |
|

uba-1
Genomic Map