Assay Details
| Gene Symbol: frk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383177.1 | 3 - 4 | 891 | 75 | ||
| NR_131592.1 | 3 - 4 | 889 | 75 | |||
| NR_131593.1 | 3 - 4 | 871 | 75 | |||
More Information
Set Membership: |
|

frk-1
Genomic Map