Assay Details
| Gene Symbol: dis-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069434.5 | NP_501835.2 | 5 - 6 | 1290 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dis-3| Gene Symbol: dis-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069434.5 | NP_501835.2 | 5 - 6 | 1290 | 71 | |
Set Membership: |
|