Assay Details
| Gene Symbol: ech-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069475.4 | NP_501876.2 | 6 - 7 | 1094 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ech-8| Gene Symbol: ech-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069475.4 | NP_501876.2 | 6 - 7 | 1094 | 100 | |
Set Membership: |
|