Assay Details
| Gene Symbol: mec-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027940.5 | NP_001023111.1 | 4 - 5 | 423 | 85 | |
| NM_001136380.4 | NP_001129852.1 | 2 - 3 | 193 | 85 | ||
| NM_001356932.4 | NP_001343802.1 | 4 | 368 | 85 | ||
| GenBank mRNA | L02877.1 | - | 4 - 5 | 390 | 85 | |
More Information
Set Membership: |
|

mec-3
Genomic Map