Assay Details
| Gene Symbol: zim-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307056.3 | NP_001293985.1 | 9 - 10 | 2729 | 72 | |
| NM_069552.3 | NP_501953.2 | 6 - 7 | 1603 | 72 | ||
| Gene Symbol: him-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383193.1 | 6 - 7 | 1600 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383193.1 transcripts.
Set Membership: |
|

zim-3
Genomic Map