Assay Details
Gene Symbol: R09H10.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069563.4 | NP_501964.2 | 4 - 5 | 392 | 86 |
Gene Symbol: F56C4.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380451.1 | NP_001367145.1 | 6 - 7 | 1440 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
