Assay Details
| Gene Symbol: mogs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083212.4 | NP_001076681.1 | 8 - 9 | 2260 | 88 | |
| NM_001083213.5 | NP_001076682.1 | 7 - 8 | 2239 | 88 | ||
| NM_001129299.4 | NP_001122771.1 | 8 - 9 | 2527 | 88 | ||
More Information
Set Membership: |
|

mogs-1
Genomic Map