Assay Details
Gene Symbol: klc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268605.3 | NP_001255534.1 | 4 - 5 | 587 | 102 | |
NM_001268606.4 | NP_001255535.1 | 3 - 4 | 550 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
