Assay Details
| Gene Symbol: cpt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047512.4 | NP_001040977.2 | 5 - 6 | 850 | 59 | |
| NM_001047513.5 | NP_001040978.2 | 3 - 4 | 340 | 59 | ||
More Information
Set Membership: |
|

cpt-2
Genomic Map