Assay Details
| Gene Symbol: gtl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069710.6 | NP_502111.3 | 30 - 31 | 5056 | 86 | |
| GenBank mRNA | AY726541.1 | - | 30 - 31 | 4986 | 86 | |
More Information
Set Membership: |
|

gtl-1
Genomic Map