Assay Details
| Gene Symbol: plx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307625.3 | NP_001294554.1 | 10 - 11 | 2094 | 64 | |
| NM_001307626.3 | NP_001294555.1 | 7 - 8 | 1527 | 64 | ||
| NM_001307627.3 | NP_001294556.1 | 7 - 8 | 1527 | 64 | ||
| NM_067617.6 | NP_500018.3 | 10 - 11 | 2094 | 64 | ||
More Information
Set Membership: |
|

plx-1
Genomic Map