Assay Details
Gene Symbol: plx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307625.3 | NP_001294554.1 | 13 - 14 | 2853 | 86 | |
NM_001307626.3 | NP_001294555.1 | 10 - 11 | 2286 | 86 | ||
NM_001307627.3 | NP_001294556.1 | 10 - 11 | 2286 | 86 | ||
NM_001307628.3 | NP_001294557.1 | 1 - 2 | 540 | 86 | ||
NM_001307629.3 | NP_001294558.1 | 1 - 2 | 540 | 86 | ||
NM_067617.6 | NP_500018.3 | 13 - 14 | 2853 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
