Assay Details
Gene Symbol: plx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307625.3 | NP_001294554.1 | 17 - 18 | 3457 | 109 | |
NM_001307626.3 | NP_001294555.1 | 14 - 15 | 2890 | 109 | ||
NM_001307627.3 | NP_001294556.1 | 14 - 15 | 2890 | 109 | ||
NM_001307628.3 | NP_001294557.1 | 5 - 6 | 1144 | 109 | ||
NM_001307629.3 | NP_001294558.1 | 5 - 6 | 1144 | 109 | ||
NM_067617.6 | NP_500018.3 | 17 - 18 | 3457 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
