Assay Details
| Gene Symbol: plx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307627.3 | NP_001294556.1 | 18 - 19 | 4333 | 87 | |
| NM_001307628.3 | NP_001294557.1 | 9 - 10 | 2587 | 87 | ||
| NM_067617.6 | NP_500018.3 | 21 - 22 | 4900 | 87 | ||
More Information
Set Membership: |
|

plx-1
Genomic Map