Assay Details
| Gene Symbol: plx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307625.3 | NP_001294554.1 | 3 - 4 | 386 | 139 | |
| NM_067617.6 | NP_500018.3 | 3 - 4 | 386 | 139 | ||
More Information
Set Membership: |
|

plx-1
Genomic Map