Assay Details
| Gene Symbol: rbm-39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383036.1 | 1 - 2 | 154 | 63 | ||
| NM_067624.5 | NP_500025.1 | 1 - 2 | 227 | 63 | ||
| NR_136395.1 | 1 - 2 | 63 | 63 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383036.1 transcripts.
Set Membership: |
|

rbm-39
Genomic Map