Assay Details
Gene Symbol: clp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067680.3 | NP_500081.2 | 5 - 6 | 1208 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
