Assay Details
| Gene Symbol: xpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067755.6 | NP_500156.2 | 9 - 10 | 3098 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

xpc-1| Gene Symbol: xpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067755.6 | NP_500156.2 | 9 - 10 | 3098 | 82 | |
Set Membership: |
|