Assay Details
| Gene Symbol: mca-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067760.5 | NP_500161.1 | 10 - 11 | 3391 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mca-2| Gene Symbol: mca-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067760.5 | NP_500161.1 | 10 - 11 | 3391 | 58 | |
Set Membership: |
|