Assay Details
| Gene Symbol: ubc-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067844.5 | NP_500245.3 | 1 - 2 | 67 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ubc-8| Gene Symbol: ubc-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067844.5 | NP_500245.3 | 1 - 2 | 67 | 78 | |
Set Membership: |
|