Assay Details
Gene Symbol: elks-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067928.4 | NP_500329.1 | 6 - 7 | 1819 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
