Assay Details
Gene Symbol: osm-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067971.4 | NP_500372.1 | 6 - 7 | 1042 | 91 | |
GenBank mRNA | AF031408.1 | - | 6 - 7 | 1042 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
