Assay Details
| Gene Symbol: cha-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028432.4 | NP_001023603.1 | 8 - 9 | 1173 | 146 | |
| GenBank mRNA | L08969.1 | - | 8 - 9 | 1182 | 146 | |
More Information
Set Membership: |
|

cha-1
Genomic Map