Assay Details
| Gene Symbol: ubc-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027987.6 | NP_001023158.1 | 3 - 4 | 295 | 77 | |
| GenBank mRNA | AF106565.1 | - | 3 - 4 | 291 | 77 | |
| Gene Symbol: F29B9.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383081.1 | 3 - 4 | 292 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383081.1 transcripts.
Set Membership: |
|

ubc-9
Genomic Map