Assay Details
| Gene Symbol: Y17G9B.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068234.7 | NP_500635.3 | 3 - 4 | 1201 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y17G9B.8| Gene Symbol: Y17G9B.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068234.7 | NP_500635.3 | 3 - 4 | 1201 | 89 | |
Set Membership: |
|