Assay Details
Gene Symbol: srz-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027936.5 | NP_001023107.2 | 2 - 3 | 310 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: srz-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027936.5 | NP_001023107.2 | 2 - 3 | 310 | 154 |
Set Membership: |
![]() ![]() ![]() ![]() |