Assay Details
| Gene Symbol: hda-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129308.4 | NP_001122780.1 | 5 - 6 | 1673 | 81 | |
| NM_068386.4 | NP_500787.1 | 5 - 6 | 1690 | 81 | ||
| NM_068387.4 | NP_500788.1 | 5 - 6 | 1690 | 81 | ||
More Information
Set Membership: |
|

hda-6
Genomic Map