Assay Details
| Gene Symbol: lsm-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068563.7 | NP_500964.1 | 3 - 4 | 462 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lsm-8| Gene Symbol: lsm-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068563.7 | NP_500964.1 | 3 - 4 | 462 | 85 | |
Set Membership: |
|