Assay Details
| Gene Symbol: acp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068581.6 | NP_500982.2 | 3 - 4 | 485 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acp-6| Gene Symbol: acp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068581.6 | NP_500982.2 | 3 - 4 | 485 | 84 | |
Set Membership: |
|