Assay Details
| Gene Symbol: fan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383106.1 | 10 - 11 | 1372 | 130 | ||
| NM_068596.5 | NP_500997.1 | 9 - 10 | 1589 | 130 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fan-1| Gene Symbol: fan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383106.1 | 10 - 11 | 1372 | 130 | ||
| NM_068596.5 | NP_500997.1 | 9 - 10 | 1589 | 130 | ||
Set Membership: |
|