Assay Details
| Gene Symbol: klp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028178.4 | NP_001023349.1 | 20 - 21 | 4778 | 141 | |
| NM_001129324.3 | NP_001122796.1 | 20 - 21 | 4703 | 141 | ||
| NM_001136394.4 | NP_001129866.1 | 3 - 4 | 907 | 141 | ||
| NM_001372925.2 | NP_001360493.1 | 19 | 4352 | 141 | ||
| Gene Symbol: T01G1.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028180.7 | NP_001023351.1 | 26 - 27 | 6011 | 141 | |
More Information
Set Membership: |
|

klp-12
Genomic Map